Learn More
QuidelOrtho ID-Micro Typing System™ Gel Test™ Cards DFS Item
Delivered with gel-filled microtubes infused with pre-measured reagents
Supplier:  QuidelOrtho MTS085014
 View more versions of this product
Catalog No. OKMTS085014
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
QuidelOrtho ID-Micro Typing System is suitable for a broad range of blood bank applications, including antibody screening and identification, ABO blood grouping and Rh phenotyping, compatibility testing, and reverse serum grouping and antigen typing, all using proven serologic methods. Gel Test cards are prefilled cards containing gel mixed with the appropriate reagent. Simply pipet, incubate, spin and read: results are ready to report, without the need for cell washing, red cell resuspension or tube shaking.
Standardization
  • Standardized methods ensure uniform results from tech-to-tech and laboratory-to-laboratory
  • Enhanced cGMP compliance is attained through standardized procedures and simplified training
Safety
  • Opportunities for error are reduced by stable and clear endpoints that provide objective, consistent results
  • Ease of Use
  • Gel testing is easy to learn, use, and teach: the ideal technology for all shifts and all levels of blood banking experience
  • Eliminates washing, shaking, and the need to add control cells
Cost-effectiveness
  • Gel testing requires fewer steps than tube testing, dramatically reducing hands-on time, with less waste and costly re-work
  • Well-balanced sensitivity and specificity means less time and fewer resources spent on clinically unimportant antibodies
Specifications
Specifications
cGMP
For broad range of Blood bank applications, Antibody screening and identification, ABO Blood grouping and Rh phenotyping, compatibility testing and reverse Serum grouping and Antigen typing
100 Cards
ID-Micro Typing System™ Buffered Gel Card
Gel-filled microtubes
Product Suggestions
Product Suggestions