missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS Stock Standard solution B for 200.8, Specpure™

Catalog No. AA40486AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40486AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40486AE Supplier Thermo Scientific Chemicals Supplier No. 040486AE
Only null left

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material ICP-MS Stock Standard solution B for 200.8
Physical Form Liquid
Concentration Matrix: 1% HNO3
MDL Number MFCD00803327
UN Number UN3264
Quantity 100 mL
Grade Specpure

RUO – Research Use Only