missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICP-MS Internal Standard solution for 200.8 & CLP ILM05.2,Specpure™
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | ICP-MS Internal Standard solution for 200.8 & CLP ILM05.2 |
| Physical Form | Liquid |
| MDL Number | MFCD03792350 |
| UN Number | UN3264 |
| Quantity | 100 mL |
| Grade | Specpure |
RUO – Research Use Only