missing translation for 'onlineSavingsMsg'
Learn More

Calibration Standard 2A For ICP/MS, SPEX CertiPrep™

Catalog No. CLMS2A
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLMS2A 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLMS2A Supplier SPEX CertiPrep Group LLC Supplier No. CLMS2A
Only null left

Includes

One bottle of Multi-Element Solution 2A and one bottle of Mercury (Hg).

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Note: The 2-pack consists of one bottle of Multi-Element Solution 2A and one bottle of Mercury (Hg). (Mercury is supplied as a separate solution due to incompatibility with other elements.)

AnalyteConcentration
Ag10μg/mL
Al10μg/mL
As10μg/mL
Ba10μg/mL
Be10μg/mL
Ca10μg/mL
Cd10μg/mL
Co10μg/mL
Cr10μg/mL
Cs10μg/mL
Cu10μg/mL
Fe10μg/mL
Ga10μg/mL
K10μg/mL
Li10μg/mL
Mg10μg/mL
Mn10μg/mL
Na10μg/mL
Ni10μg/mL
Pb10μg/mL
Rb10μg/mL
Se10μg/mL
Sr10μg/mL
Tl10μg/mL
U10μg/mL
V10μg/mL
Zn10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Multi-Element Solution 2A
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Solvent or Matrix 5% HNO3
Quantity 125 mL
Grade ICP/ICP-MS
Concentration or Composition (by Analyte or Components) 10mg/L
Show More Show Less

Product intended for laboratory use only