missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calibration Standard 2 without Mercury For ICP/MS, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC CLMS2N
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 10ÎĽg/mL |
Al | 10ÎĽg/mL |
As | 10ÎĽg/mL |
Ba | 10ÎĽg/mL |
Be | 10ÎĽg/mL |
Bi | 10ÎĽg/mL |
Ca | 10ÎĽg/mL |
Cd | 10ÎĽg/mL |
Co | 10ÎĽg/mL |
Cr | 10ÎĽg/mL |
Cs | 10ÎĽg/mL |
Cu | 10ÎĽg/mL |
Fe | 10ÎĽg/mL |
Ga | 10ÎĽg/mL |
In | 10ÎĽg/mL |
K | 10ÎĽg/mL |
Li | 10ÎĽg/mL |
Mg | 10ÎĽg/mL |
Mn | 10ÎĽg/mL |
Na | 10ÎĽg/mL |
Ni | 10ÎĽg/mL |
Pb | 10ÎĽg/mL |
Rb | 10ÎĽg/mL |
Se | 10ÎĽg/mL |
Sr | 10ÎĽg/mL |
Tl | 10ÎĽg/mL |
U | 10ÎĽg/mL |
V | 10ÎĽg/mL |
Zn | 10ÎĽg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Multi-Element Solution 2 without Mercury | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
ICP/ICP-MS Standard | |
125 mL | |
10mg/L |
Triple-leached LDPE bottle | |
ICP-MS | |
Standard Solution | |
5% HNO3 | |
ICP/ICP-MS |
Product intended for laboratory use only