missing translation for 'onlineSavingsMsg'
Learn More

Icariin 96.0+%, TCI America™

Catalog No. i08621g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
I08621G 1 g
I0862200MG 200 mg
2 options

Catalog No. I08621G

Supplier: TCI America I08621G

Only null left

Chemical Identifiers

CAS 489-32-7
Molecular Formula C33H40O15
Molecular Weight (g/mol) 676.668
MDL Number MFCD00210516
InChI Key TZJALUIVHRYQQB-JOJMVVPMSA-N
PubChem CID 131801625
IUPAC Name 5-hydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-enyl)-7-[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxychromen-4-one
SMILES CC1C(C(C(C(O1)OC2=C(OC3=C(C2=O)C(=CC(=C3CC=C(C)C)OC4C(C(C(C(O4)CO)O)O)O)O)C5=CC=C(C=C5)OC)O)O)O

Specifications

Melting Point 237°C
Color Yellow
Quantity 1 g
Formula Weight 676.67
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Icariin
RTECSNumber DJ2980500
TSCA No
Recommended Storage Refrigerator