missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibutilide Hemifumarate 98.0+%, TCI America™
$339.25 - $339.25
Chemical Identifiers
| CAS | 122647-32-9 |
|---|---|
| Molecular Formula | C44H76N4O10S2 |
| Molecular Weight (g/mol) | 885.23 |
| MDL Number | MFCD01715410 |
| InChI Key | PCIOHQNIRPWFMV-WXXKFALUNA-N |
| Synonym | ibutilide fumarate, corvert, ibutilide fumarate usan, ibutilide hemifumarate, ibutilide hemifumarate salt, corvert tn, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfoanilide e-2-butenedioate 2:1, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfonanilide fumarate 2:1 salt, +--n-4-4-ethylheptylamino-1-hydroxybutyl phenyl methanesulfonamide e-butenedioate |
| PubChem CID | 5281065 |
| IUPAC Name | (2E)-but-2-enedioic acid; bis(N-(4-{4-[ethyl(heptyl)amino]-1-hydroxybutyl}phenyl)methanesulfonamide) |
| SMILES | OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I0946100MG
|
TCI America
I0946100MG |
100 mg |
Each for $339.25
|
|
|||||
Chemical Identifiers
| 122647-32-9 | |
| 885.23 | |
| PCIOHQNIRPWFMV-WXXKFALUNA-N | |
| 5281065 | |
| OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1 |
| C44H76N4O10S2 | |
| MFCD01715410 | |
| ibutilide fumarate, corvert, ibutilide fumarate usan, ibutilide hemifumarate, ibutilide hemifumarate salt, corvert tn, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfoanilide e-2-butenedioate 2:1, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfonanilide fumarate 2:1 salt, +--n-4-4-ethylheptylamino-1-hydroxybutyl phenyl methanesulfonamide e-butenedioate | |
| (2E)-but-2-enedioic acid; bis(N-(4-{4-[ethyl(heptyl)amino]-1-hydroxybutyl}phenyl)methanesulfonamide) |
Specifications
| 122647-32-9 | |
| C44H76N4O10S2 | |
| 100 mg | |
| PCIOHQNIRPWFMV-WXXKFALUNA-N | |
| (2E)-but-2-enedioic acid; bis(N-(4-{4-[ethyl(heptyl)amino]-1-hydroxybutyl}phenyl)methanesulfonamide) | |
| 5281065 | |
| ≥98.0% (HPLC,N) | |
| Ibutilide Hemifumarate |
| White | |
| MFCD01715410 | |
| ibutilide fumarate, corvert, ibutilide fumarate usan, ibutilide hemifumarate, ibutilide hemifumarate salt, corvert tn, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfoanilide e-2-butenedioate 2:1, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfonanilide fumarate 2:1 salt, +--n-4-4-ethylheptylamino-1-hydroxybutyl phenyl methanesulfonamide e-butenedioate | |
| OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1 | |
| 885.23 | |
| 442.62 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : PB0475700
TSCA : No