missing translation for 'onlineSavingsMsg'
Learn More

Ibutilide Hemifumarate 98.0+%, TCI America™

Catalog No. I0946100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0946100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0946100MG Supplier TCI America Supplier No. I0946100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122647-32-9
Molecular Formula C44H76N4O10S2
Molecular Weight (g/mol) 885.23
MDL Number MFCD01715410
InChI Key PCIOHQNIRPWFMV-WXXKFALUNA-N
Synonym ibutilide fumarate, corvert, ibutilide fumarate usan, ibutilide hemifumarate, ibutilide hemifumarate salt, corvert tn, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfoanilide e-2-butenedioate 2:1, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfonanilide fumarate 2:1 salt, +--n-4-4-ethylheptylamino-1-hydroxybutyl phenyl methanesulfonamide e-butenedioate
PubChem CID 5281065
IUPAC Name (2E)-but-2-enedioic acid; bis(N-(4-{4-[ethyl(heptyl)amino]-1-hydroxybutyl}phenyl)methanesulfonamide)
SMILES OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1

Specifications

Color White
Quantity 100 mg
Formula Weight 442.62
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ibutilide Hemifumarate
RTECSNumber PB0475700
TSCA No