missing translation for 'onlineSavingsMsg'
Learn More

Medisca Ibuprofen USP

Catalog No. 14185400
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185400 25 g
14185384 100 g
14185449 1 kg
14185164 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14185400 Supplier Medisca Supplier No. 029904
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ibuprofen USP is a white to off-white, crystalline powder, having a slight, characteristic odor.

Ibuprofen USP is a white to off-white, crystalline powder, having a slight, characteristic odor.

Specifications

Chemical Name or Material Ibuprofen USP
Packaging Jar 100ml, White, HDPE, Round, 38-400, TRIVENI-DB, CT
CAS 15687-27-1
Type Drug Standard
Molecular Weight (g/mol) 206.28
Quantity 25 g
Percent Purity 97 to 103 %
Grade USP