missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Ibuprofen USP
Description
Ibuprofen USP is a white to off-white, crystalline powder, having a slight, characteristic odor.
Specifications
Specifications
| Chemical Name or Material | Ibuprofen USP |
| Packaging | Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT |
| CAS | 15687-27-1 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 206.28 |
| Quantity | 100 g |
| Percent Purity | 97 to 103 % |
| Grade | USP |