missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Impurity F Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015080
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015080 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015080 Supplier Merck Emd Millipore Supplier No. PHR196430MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ibuprofen Impurity F is an impurity of the drug, ibuprofen. Ibuprofen is a non-steroidal anti-inflammatory drug (NSAID) that is effective against pain, inflammation and fever. It is deemed as a safe drug on account of its reversible inhibitory effects on cyclooxygenase (COX) enzyme along with its unique pharmacokinetics.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 65322-85-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C13H18O2
Synonym 3-(4-Isobutylphenyl)propionic acid; 3-[4-(2-METHYLPROPYL)PHENYL] PROPANOIC ACID
Type Certified Reference Material
Quantity 30 mg
Formula Weight 206.28
Grade Certified Reference Material