missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Impurity B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111003451
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111003451 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111003451 Supplier Merck Emd Millipore Supplier No. PHR193420MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C13H17O2
Synonym Ibuprofen Impurity B Sodium Salt; (2 RS)-2- (4-BUTYLPHENYL)PROPANOIC ACID SODIUM SALT
Type Certified Reference Material
Quantity 20 mg
Formula Weight 205.27
Grade Certified Reference Material