missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibuprofen 1,3-Butylene Glycol Esters Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C17H26O3 |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 278.39 |
| Grade | Certified Reference Material |