missing translation for 'onlineSavingsMsg'
Learn More

Ibudilast 98.0+%, TCI America™

Catalog No. p-7131572
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I074020MG 20 mg
I0740100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I074020MG Supplier TCI America Supplier No. I074020MG
Only null left

Chemical Identifiers

CAS 50847-11-5
Molecular Formula C14H18N2O
Molecular Weight (g/mol) 230.311
MDL Number MFCD00864808
InChI Key ZJVFLBOZORBYFE-UHFFFAOYSA-N
Synonym ibudilast, ketas, ibudilastum, ke tas, ibudilastum latin, ibudilast inn:jan, eyevinal, unii-m0tth61xc5, ketas tn, 3-isobutyryl-2-isopropylpyrazolo 1,5-a pyridine
PubChem CID 3671
IUPAC Name 2-methyl-1-(2-propan-2-ylpyrazolo[1,5-a]pyridin-3-yl)propan-1-one
SMILES CC(C)C1=NN2C=CC=CC2=C1C(=O)C(C)C

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 175°C
Quantity 20 mg
Formula Weight 230.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ibudilast
RTECSNumber UR0711200
TSCA No
Recommended Storage Refrigerator