accessibility menu, dialog, popup

UserName

CAS RN 50847-11-5

IUPAC Name: 2-methyl-1-[2-(propan-2-yl)pyrazolo[1,5-a]pyridin-3-yl]propan-1-one
Synonyms: ibudilast I0157_sigma ibudilast (jan/inn) ibudilastum unii-M0TTH61XC5 ke tas tocris-1694 eyevinal ketas (TN) ketas
Molecular Weight (g/mol): 230.31
Molecular Formula: C14H18N2O
InChi Key: ZJVFLBOZORBYFE-UHFFFAOYSA-N
SMILES: CC(C)C(=O)C1=C2C=CC=CN2N=C1C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results