missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBright™ System Tray Adapter for E-Gel™ Agarose Gels

Catalog No. p-7227791
Encompass
Change view
Click to view available options
Product Type:
E-Gel Tray Adapter, 11-/22-well
E-Gel Tray Adapter, 48-/96-well
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
A56600 E-Gel Tray Adapter, 11-/22-well
A56599 E-Gel Tray Adapter, 48-/96-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A56600 Supplier Invitrogen™ Supplier No. A56600
Only null left

Imaging accessories designed to optimize the image capture of Invitrogen E-Gel Agarose Gels.

Invitrogen iBright Tray Adapters for E-Gel Agarose Gels are imaging accessories designed to optimize the image capture of Invitrogen E-Gel Agarose Gels.

Features of Invitrogen iBright Tray Adapters for E-Gel Agarose Gels include:
• The bottom component centers the E-Gel agarose gels on the iBright transilluminator box facilitating optimized zoom
• The top component covers labels and bar codes on the E-Gel cassette to reduce stray light during image capture and improve sample signal optimization
• Compatible with all iBright models (CL750, CL1000, CL1500, FL1000, and FL1500)
• Compatible with 11- to 96-well E-Gel agarose (Cat. No. A56600 for 11-/22-well, Cat. No. A56599 for 48-/96-well)

Specifications

Product Line iBright
For Use With (Application) Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging
Quantity 1 Adaptor
Shipping Condition Room Temperature
Product Type E-Gel Tray Adapter, 11-/22-well