missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBright™ System Contrast Tray

Catalog No. A56597
Encompass
Change view
Click to view available options
Quantity:
1 Tray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A56597 1 Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A56597 Supplier Invitrogen™ Supplier No. A56597

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This iBright System Contrast Tray is an iBright imaging system accessory designed to improve the imaging of visible protein gels.

  • This iBright System Contrast Tray is an iBright imaging system accessory designed to improve the imaging of visible protein gels
  • The dual layer tray is designed to create optimal lighting conditions when used with the green LED transilluminator on the iBright imaging system, improving the contrast and lowering the overall background to capture high-quality visible-stain images
  • Compatible with all iBright models (CL750, CL1000, CL1500, FL1000, and FL1500)
  • Provides a contrasting background when imaging protein gels stained with colorimetric stains, such as Coomassie blue and silver stains
  • This tray also improves conditions when samples contain dyes that auto-fluoresce under green LED light
  • Recommended for transillumination of crystal violet-stained colony plates
  • A replacement for the White Transilluminator Screen (Cat. No. A33828)

Specifications

Product Line iBright
For Use With (Application) Chemiluminescent Western Blot Imaging, DNA Gel Imaging, Protein Gel Imaging
Product Type Contrast Tray
Quantity 1 Tray
Shipping Condition Room Temperature
What dyes and stains are supported by the iBright Imaging Systems?

The iBright Imaging Systems support a wide range of dyes, stains, and fluorophores from the visible to NIR wavelength range.

For more information on compatible reagents and applications, visit this page.

To learn more about the iBright system-supported fluorophores and installed filters and light sources, visit Fluorescence SpectraViewer.

How large is the sample imaging area on the iBright Imaging Systems?

The imaging area of the iBright Imaging system is 22.5 x 18.0 cm. This gives ample space for imaging 4 mini blots (or 2 midi blots) simultaneously.

For more information on iBright features and capabilities, visit this page.

What sample types can I image with the iBright CL750 Imaging System?

The iBright CL750 Imaging System supports imaging of chemiluminescent western blots, colorimetric and fluorescent stained gels, and nucleic acid gels. It can also be used to image GFP- and crystal violet-stained colony plates.

For additional information about these applications or to learn more about the three iBright model capabilities, visit this page.

To speak with a representative about the iBright imager or request a quote or demo, please fill out this form.

What sample types can I image with the iBright CL1500 Imaging System?

The iBright CL1500 Imaging System supports a wide range of applications including the following:

Protein gel: colorimetric staining of gels (e.g., Coomassie, silver) and membranes (e.g., Ponceau S, Thermo Scientific Pierce Reversible Protein Stain), fluorescent staining of gels (e.g., Invitrogen SYPRO Ruby stain)

Nucleic acid gel: ethidium bromide and Invitrogen SYBR stains; imaging of E-Gels and hand pour gels is supported

Chemiluminescent western blot: chemiluminescence using all popular HRP and AP substrates (e.g., Thermo Scientific SuperSignal and Invitrogen WesternBreeze substrates)

Additional applications: colony counting for GFP, crystal violet, and blue/white colonies; TLC plates, and leaf sections

For additional information about these applications or to learn more about the three iBright model capabilities, visit this page.

To speak with a representative about the iBright imager or request a quote or demo, please fill out this form.

What sample types can I image with the iBright FL1500 Imaging System?

The iBright FL1500 Imaging System provides maximum application support including up to 4-channel multiplex western blot imaging. See the list below for more details:

Protein gel: colorimetric staining of gels (e.g., Coomassie, silver) and membranes (e.g., Ponceau S, Thermo Scientific Pierce Reversible Protein Stain), fluorescent staining of gels (e.g., Invitrogen SYPRO Ruby stain)

Nucleic acid gel: ethidium bromide and Invitrogen SYBR stains; imaging of E-Gels and hand-poured gels is supported

Chemiluminescent western blot: chemiluminescence using all popular HRP and AP substrates (e.g., Thermo Scientific SuperSignal and Invitrogen WesternBreeze substrates)

Fluorescent western blot: fluorescence with popular RGB (visible range) and near-IR fluorophores (e.g., Invitrogen Alexa Fluor and Alexa Fluor Plus conjugates)

Additional applications: colony counting for GFP, crystal violet, and blue/white colonies; TLC plates, and leaf sections

For additional information about these applications or to learn more about the three iBright model capabilities, visit this page.

To speak with a representative about the iBright imager or request a quote or demo, please fill out this form.

Is the iBright System Contrast Tray (Cat. No. A56597) included with purchase of an iBright imager?

The iBright System Contrast Tray is included with the purchase of a current iBright Imaging System, inclusive of the CL750, CL1500, and FL1500 iBright Imaging Systems. For additional information on iBright features and capabilities, visit the iBright Imaging Systems page.

Which iBright imager models are compatible with the iBright System Contrast Tray (Cat. No. A56597)?

All iBright imager models are compatible with the orange iBright System Contrast Tray (Cat. No. A56597), inclusive of the CL1000, FL1000, CL750, CL1500, and FL1500 iBright imager models. For more information on compatible reagents and applications, visit the iBright Imaging Systems Compatible Reagents page.

What sample types can I image with the iBright System Contrast Tray (Cat. No. A56597)?

We recommend using the iBright System Contrast Tray (Cat. No. A56597) for imaging all visible protein gel samples such as those stained with Coomassie Blue or Silver Stain. For more information on compatible reagents and applications, visit the iBright Imaging Systems Compatible Reagents page.

What is the best way to image Ponceau staining with an iBright Imaging System?

The recommended imaging method depends on the iBright imager model being used. For iBright FL1500 and CL1500 Imaging Systems, we recommend imaging Ponceau-stained samples with the Ponceau application that can be found in Universal mode. For the iBright CL750 Imaging System, we recommend imaging Ponceau-stained samples in Protein gel mode.

We always recommend imaging with the orange iBright System Contrast Tray (Cat. No. A56597), and if needed, adjust the focus or zoom to get bands as sharp as possible. Better quality pictures are obtained when the membrane is wet.


For Research Use Only. Not for use in diagnostic procedures.