missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBlot™ Transfer Stack, nitrocellulose, regular size
GREENER_CHOICE

iBlot Transfer Stacks are used to transfer proteins.

Supplier:  Invitrogen™ IB301001

Encompass

 View more versions of this product

Catalog No. IB301001


Only null left

Description

Description

The iBlotTransfer Stacks are disposable stacks that come in three varieties for different applications: iBlotGel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins; iBlot DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlotTransfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.

Nitrocellulose membrane

  • Composed of 100% pure nitrocellulose to provide high-quality transfer
  • Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
  • Bind proteins due to hydrophobic and electrostatic interactions
  • Pore size: 0.2μm
  • Protein binding capacity: 209μg/cm2

 

Transfer stack sizes

  • The regular size (13 x 8.3cm) is used for all Novex midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels
  • The mini size (8 x 8cm) is used for all Novex mini-gels (or equivalent)

 

Proteins, Expression, Isolation and Analysis, Western Blotting

Specifications

Chemical Identifiers

13.5 cm
Transfer Stack
0.2 μm
iBlot
8 cm
Nitrocellulose
Regular, 13.5 cm x 8 cm

Specifications

13.5 cm
10 sets
Nitrocellulose
10 sets of transfer stacks in 1 box. Store at room temperature.
Regular, 13.5 cm x 8 cm
10 Blots
8 cm
Transfer Stack
0.2 μm
Room Temperature
iBlot
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.