Learn More
Invitrogen™ iBind™ Flex Cards DFS Item
Card for use with iBind™ Flex Western Blot Device.
Supplier:  Invitrogen™ SLF2010X4
 View more versions of this product
Catalog No. SLF2010X4
Only null left
Description
Description
  • Glass fiber backed card with filer paper stack at base
  • Quantity:  40 Cards (4 boxes of 10)
Specifications
Specifications
Store at room temperature.
Chromogenic, Chemiluminescence, Fluorescence
iBind Flex Western Device
iBind Flex
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots/Card
40 Cards (10 x 4)
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Western Blot
Nitrocellulose, PVDF
∼2.5 hr.
Card
Room Temperature
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.