missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ iBind™ Flex Cards DFS Item

Card for use with iBind™ Flex Western Blot Device.

$479.00 - $1566.00

Specifications

Capacity 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Detection Method Chromogenic, Chemiluminescence, Fluorescence
For Use With (Application) Western Blot
For Use With (Equipment) iBind Flex Western Device
Membrane Compatibility Nitrocellulose, PVDF
View More Specs

Products 2
Catalog Number Mfr. No. Type Quantity Price Quantity  
Catalog Number Mfr. No. Type Quantity Price Quantity  
SLF2010X4 DFS Item
View Documents
Invitrogen™
SLF2010X4
Card 40 Cards (10 x 4)
Pack of 4 Each for $1,566.00
Only null left
 
SLF2010 DFS Item
View Documents
Invitrogen™
SLF2010
Card 10 Cards
Each for $479.00
Only null left
 
Description

Description

  • Glass fiber backed card with filer paper stack at base
  • 10 cards per box
Specifications

Specifications

1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Western Blot
Nitrocellulose, PVDF
∼2.5 hr.
Card
Chromogenic, Chemiluminescence, Fluorescence
iBind Flex Western Device
iBind Flex
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots/Card
Room Temperature
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.