missing translation for 'onlineSavingsMsg'
Learn More

Mirus Bio™ TransIT™-Keratinocyte Transfection Reagent

Catalog No. MIR2804
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
5 mL
10 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR2804 0.4 mL
MIR2800 1 mL
MIR2805 5 mL
MIR2806 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MIR2804 Supplier Mirus Bio™ Supplier No. MIR2804
Only null left

Includes

TransIT-Keratinocyte transfection reagent, supplied as a ready-to-use solution.

TransIT™-Keratinocyte Transfection Reagent designed for highly efficient, low-toxicity DNA transfection in primary keratinocytes. Transfection does not required medium changes and can be carried out in serum-containing medium.

  • TransIT-Keratinocyte transfection reagent, supplied as a ready-to-use solution.
  • Optimized transfection performance in keratinocyte cells.
  • Low cellular toxicity maintains cell density and reduces experimental bias.
  • Allows 20 to 30% transfection efficiency in keratinocytes.
  • Concentration: 1.37 mg/mL

Specifications

Cell Type Mammalian
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Concentration 1.37 mg/mL
For Use With (Application) Highly efficient DNA or RNA transfection with minimal cellular toxicity, specifically for primary keratinocytes
Product Type Transfection Reagent
Quantity 0.4 mL
Sterility Sterile