missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Taq Buffers (10X)

Catalog No. FERB55
Change view
Click to view available options
Chemical Name or Material:
Taq Buffer with (NH4)2SO4
Taq Buffer with (NH4)2SO4 and MgCl2
Taq Buffer with KCl
Taq Buffer with KCl and MgCl2
Taq Buffer without Detergent
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
FERB55 Taq Buffer without Detergent
FERB38 Taq Buffer with KCl
FERB16 Taq Buffer with KCl and MgCl2
FERB33 Taq Buffer with (NH4)2SO4
FERB34 Taq Buffer with (NH4)2SO4 and MgCl2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. FERB55 Supplier Thermo Scientific™ Supplier No. B55

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.

Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA polymerases (both recombinant and native). They are available in different compositions and also without detergents.

10X Taq Buffer without Detergent

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl

10X Taq Buffer with KCl and 15 mM MgCl2

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl
  • 15 mM MgCl2
  • Detergent

10X Taq Buffer with (NH4)2SO4

  • 750 mM Tris-HCl (pH 8.8 at 25°C)
  • 200 mM (NH4)2SO4
  • 0.1% (v/v) Tween 20

10X Taq Buffer with (NH4)2SO4 and 20 mM MgCl2

  • 750 mM Tris-HCl (pH 8.8 at 25°C)
  • 200 mM (NH4)2SO4
  • 0.1% (v/v) Tween 20
  • 20 mM MgCl2

10X Taq Buffer with KCl

  • 100 mM Tris-HCl (pH 8.8 at 25°C)
  • 500 mM KCl
  • Detergent

Specifications

Chemical Name or Material Taq Buffer without Detergent
Concentration 10X
For Use With (Application) Standard PCR
Physical Form Liquid
Quantity 4 x 1.25 mL
Includes • 10X Taq Buffer without Detergent, 4 x 1.25 mL

Store at -20°C.

For Research Use Only. Not for use in diagnostic procedures.