missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ I-PER™ Insect Cell Protein Extraction Reagent

Catalog No. PI89802
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI89802 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI89802 Supplier Thermo Scientific™ Supplier No. 89802

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Extract soluble protein from baculovirus-infected insect cells grown in suspension or monolayer culture with this gentle and effective lysis reagent.

Thermo Scientific™ I-PER Insect Cell Protein Extraction Reagent enables gentle and effective extraction of soluble protein from baculovirus-infected insect cells grown in suspension or monolayer culture.

The baculovirus insect cell expression system is an efficient and popular system for the production of recombinant (eukaryotic) proteins in cell culture. Proteins expressed in baculoviral systems can be used for structural analyses, biochemical assays and a variety of other applications. I-PER Reagent maintains functionality of extracted proteins and is directly compatible with downstream applications such as protein assays, Western blotting and 6xHis-tagged protein purification.

Highlights:

  • Gentle extraction – optimized, mild non-ionic detergent provides maximum extraction of soluble proteins from insect cells

  • Effective – provides better protein extraction than sonication

  • Compatible – downstream compatibility with Western blotting, 6xHis-tagged protein purification, protein assays and ion-exchange chromatography

  • Flexible – useful for protein extraction from suspended or adherent cultured insect cells

Specifications

Quantity 250 mL
Format Liquid
Product Type Reagent
Volume (Metric) 250 mL
Content And Storage Upon receipt store at 4°C.
Product Line I-PER
Can you provide the shelf-life for the I-PER Insect Cell Protein Extraction Reagent?

The I-PER Insect Cell Protein Extraction Reagent is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.