missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mirus Bio™ flashBAC™ Baculovirus Expression System
Description
The original flashBAC expression vector. flashBAC expression systems are a streamlined platform for the production of recombinant baculoviruses. Genetic optimization of the Autographa californica nucleopolyhedrovirus (AcMNPV) genome yields recombinant virus in a single step and eliminates the need for separation of recombinant and parental virus by plaque purification. Protein expression and secretion is improved by deletion of the chitinase gene from the AcMNPV genome. Optimal for expression of most nuclear or cytoplasmic proteins.
- Single-step baculovirus generation in insect cells, no plaque purification required
- Ready to use with most transfer vectors, including BacMam
- High recombinant protein yields
Warranty
1 Year
Specifications
Specifications
| Content And Storage | Store flashBAC™ DNA at 4°C and control plasmid at -20°C |
| Cell Type | Insect Cells |
| Format | Kit |
| Sample Type | Baculovirus DNA, Plasmid DNA |
| Promoter | None (Promoterless) |
| Transfection Technique | Chemical Transfection |
| For Use With (Application) | Transfection |
| No. of Reactions | 24 |
| Product Type | Transfection Reagent |
| Sterility | Sterile |