missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ I-Block™ Protein-Based Blocking Reagent DFS Item

Highly purified Casein-based blocking reagent. Thermo Scientific I-Block Protein-Based Blocking Reagent provides superior blocking compared to both dried milk and BSA.

Supplier:  Invitrogen™ T2015

Catalog No. T2015


Only null left
Explore available promotions

Description

Description

  • Biotin-free
  • Tested in assays using Tropix substrates and alkaline phosphatase conjugates
  • Useful as blocking reagent in membrane-based and immunoassay applications
  • Suggested working
  • concentration is 0.2% (w/v) for detection of nucleic acids on neutral or positively-charged nylon membranes and for immunoassays and protein detection on membranes (nitrocellulose, PVDF, or neutral nylon)
  • Concentration of 3% is recommended for protein detection on positively-charged nylon membrane (such as TropilonPlus™ membrane)

Preparation:
Prepared in either Tris- or phosphate-buffered saline buffer with heating (40° to 50°C).

Applications:
Can be used with Western-Light™, Western-Light Plus™, Western-Star™, ELISA-Light™, Southern-Light™, and Southern-Star™ Detection Systems.

Specifications

Specifications

Reagent
Western-Light™ Detection System, Western-Star™ Detection System, Southern-Star™ Detection System, ELISA-Light™ Detection System, Southern-Light™ Detection System, Western-Light Plus™ Detection System
I-Block, NovaBright
30 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.