missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BacLight™ RedoxSensor™ CTC Vitality Kit

Catalog No. 361045886
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361045886 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361045886 Supplier Invitrogen™ Supplier No. B34956

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The BacLight™ RedoxSensor™ CTC Vitality Kit provides effective reagents for evaluating bacterial cell health and vitality that can withstand fixation procedures.

The BacLight™ RedoxSensor™ CTC Vitality Kit provides effective reagents for evaluating bacterial cell health and vitality that can withstand fixation procedures. Briefly, healthy cells respiring via the electron transport chain will absorb and reduce CTC into an insoluble, red fluorescent formazan product. Cells not respiring or respiring at slower rates will reduce less CTC and consequently produce less fluorescent product, giving a semi-quantitative estimate of healthy vs. unhealthy bacteria. Green- and blue-fluorescent nucleic acid stains are included as counterstains to assist differentiation of cells from debris and calculating total cell numbers.

Specifications

Cell Type Bacteria
Content And Storage • 5-cyano-2,3-ditolyl tetrazolium chloride (CTC), 5 x 15 mg vials
• SYTO™ 24 green (100 μL in DMSO), 1 vial
• DAPI (5 mg/mL solution in water), 100 μL

Store in freezer (-5°C to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type CTC Vitality Kit
Dye Type Other Label(s) or Dye(s)
Format Tube(s), Slide(s)
Product Line BacLight, RedoxSensor
Quantity 1 Kit
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Emission Wavelength Range 488
Show More Show Less
What bacterial parameters can I look at by flow cytometry?

You can stain bacteria with a general stain such as BacLight Green Bacterial Stain (Cat. No. B35000) or BacLight Red Bacterial Stain (Cat. No. B35001). You can look at gram character (Cat. No. L7005), cell viability (Cat. Nos. L7007, L7012, and L13152), cell count (Cat. Nos. L34856 and B7277), and cell vitality. Cell vitality can be measured by membrane potential (Cat. No. B34950) or by metabolism (Cat. Nos. B34954 and B34956).

For Research Use Only. Not for use in diagnostic procedures.