missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ BacLight™ Green Bacterial Stain

Catalog No. B35000
Change view
Click to view available options
Quantity:
20 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35000 20 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B35000 Supplier Molecular Probes™ Supplier No. B35000

BacLight™ Green Bacterial Stain

The BacLight Green (Cat. no. B-35000) and BacLight Red (Cat. no. B-35001) bacterial stains are fluorescent labeling reagents for detecting and monitoring bacteria. These two dyes are not nucleic acid stains. Bacteria stained with the BacLight Green and BacLight Red bacterial stains exhibit bright green and red fluorescence (absorption/emission ∼480/516 and ∼581/644 nm, respectively), and can be resolved using the appropriate flow cytometric channels or fluorescence microscopy filters. The BacLight bacterial stains are compatible with formaldehyde or alcohol fixation methods.

Specifications

Cell Type Bacteria
Content And Storage Contains 20 vials of BacLight™ green bacterial stain (50 ug per vial). Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Bacterial Stain
Dye Type Other Label(s) or Dye(s)
Form Solid
Format Tube(s), Slide(s)
Product Line BacLight
Quantity 20 x 50 μg
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Emission Wavelength Range 480/ 561
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.