Learn More
BUCHI I-180 Controller for Rotavapor R-80
The I-180 is an easy-to-use interface for conveniently regulating your vacuum automatically. Increase control of the evaporation process by setting the desired vacuum and accessing other modes of operation.
$3887.80 - $6199.22
Specifications
Type Controller Interface
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Includes Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Includes Price Quantity  
01-672-992
View Documents
BUCHI
11I1802
Rotavapor R-80 V-80/180 Power Pack
Each for $6,199.22
Only null left
01-672-993
View Documents
BUCHI
11I1801
Rotavapor R-80 V-80/180 Communication Cable, Holder
Each for $3,887.80
Only null left
01-672-994
View Documents
BUCHI
11I1800
Rotavapor R-80 V-80/180 House Vacuum Valve Unit, Woulff Bottle, Communication Cable, Tubing
Each for $6,087.85
Only null left
01-672-991
View Documents
BUCHI
11I1803
Rotavapor R-80 I-100 Adapter Cable for I-180, I-100 Power Supply
Each for $3,966.61
Only null left
Description
Description
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
Specifications
Specifications
Controller Interface