Learn More
BUCHI I-180 Controller for Rotavapor R-80
Catalog No. 01672992
Change view
Click to view available options
For Use With (Equipment):
Rotavapor R-80 I-100
Rotavapor R-80 V-80/180
Rotavapor R-80 V-80/180 House Vacuum
Includes:
Adapter Cable for I-180, I-100 Power Supply
Communication Cable, Holder
Power Pack
Valve Unit, Woulff Bottle, Communication Cable, Tubing
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Includes
01672992 Rotavapor R-80 V-80/180 Power Pack
01672993 Rotavapor R-80 V-80/180 Communication Cable, Holder
01672994 Rotavapor R-80 V-80/180 House Vacuum Valve Unit, Woulff Bottle, Communication Cable, Tubing
01672991 Rotavapor R-80 I-100 Adapter Cable for I-180, I-100 Power Supply
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01672992 Supplier BUCHI Supplier No. 11I1802
The I-180 is an easy-to-use interface for conveniently regulating your vacuum automatically. Increase control of the evaporation process by setting the desired vacuum and accessing other modes of operation.
  • Functional design fulfills essential needs of vacuum regulation
  • Saves resources thanks to control of vacuum pump
  • High longevity due to robust and durable design
  • User-friendly and intuitive handling due to self-explaining user interface
  • Easy and accurate vacuum regulation
  • Relevant information is displayed at a glance
  • High reliability due to centralized control of major process parameters
  • Full process control thanks to parameter monitoring
  • Proven technology for major applications
Specifications
Type Controller Interface
For Use With (Equipment) Rotavapor R-80 V-80/180
Includes Power Pack