missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STERIS Hypochlorite WFI Sterile Solution
Oxidizing cleaners can be extremely effective when removing tough soils and residue from hard surfaces, even the most critical surfaces
Supplier: STERIS 1S0877WR
Description
- Designed to remove tough soil and residue from surfaces
- Made with United States Pharmacopeia (USP) Water For Injection and Sodium Hypochlorite
- Filtered, and filled in an ISO Class 5 aseptic environment to minimize the potential of particles being introduced during use
- Packages are double bagged for easy entry into controlled environments
- Includes an extensive documentation package to meet validation objectives
- Each lot documented by a Certificate of Analysis including sterility results
- Supported by the STERIS Technical Services team for individualized application and validation assistance
- These products are not registered as disinfectants by the US EPA
Specifications
| Bleach | |
| Sodium Hypochlorite, USP Water For Injection (WFI) | |
| Trigger Spray Bottle | |
| Removing Tough Soil and Residue From Surfaces | |
| 22 oz. |
| Sterile | |
| 0.525% | |
| Filtered | |
| 12/Cs. |