Learn More
Vikan™ Hygienic Wall Bracket, Grip Band Module
This grip band module is designed for storing cleaning tools without a hanging hole.
$32.21 - $32.21
Specifications
| Product Type | Wall Bracket |
|---|---|
| Material | Polypropylene |
| Height (English) | 1.9 in. |
| Length (English) | 3.2 in. |
| Weight (English) | 0.18 lb. |
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
|
17228024
|
Vikan™
10131 |
Pink |
Each for $32.21
|
|
|||||
|
17228025
|
Vikan™
REM10132 |
Green |
Each for $32.21
|
|
|||||
|
17228026
|
Vikan™
10133 |
Blue |
Each for $32.21
|
|
|||||
|
17228027
|
Vikan™
10134 |
Red |
Each for $32.21
|
|
|||||
|
17228028
|
Vikan™
10135 |
White |
Each for $32.21
|
|
|||||
|
17228029
|
Vikan™
10136 |
Yellow |
Each for $32.21
|
|
|||||
|
17228033
|
Vikan™
101366 |
Brown |
Each for $32.21
|
|
|||||
|
17228030
|
Vikan™
10137 |
Orange |
Each for $32.21
|
|
|||||
|
17228034
|
Vikan™
101377 |
Lime |
Each for $32.21
|
|
|||||
|
17228031
|
Vikan™
10138 |
Purple |
Each for $32.21
|
|
|||||
|
17228035
|
Vikan™
REM101388 |
Gray |
Each for $32.21
|
|
|||||
|
17228032
|
Vikan™
10139 |
Black |
Each for $32.21
|
|
|||||
Description
- Slide the grip band module onto the supplied double-base/ spacer from either side
- This grip module can hold products with a diamter of 28 to 33 mm
- Easy to dissemble for cleaning or replacement
Specifications
| Wall Bracket | |
| 1.9 in. | |
| 0.18 lb. |
| Polypropylene | |
| 3.2 in. | |
| 3.4 in. |