missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ HyFlex™ INTERCEPT™ Cut Resistant Medium Duty Industrial Sleeves with Thumbslot

Made of black yarn and are silicone and latex free for friendly paint and finish process

$484.89 - $484.89

Specifications

Product Type Cut-resistant Sleeve
Material Ansell INTERCEPT™ Technology
Color Dark Gray
Sterility Non-sterile
Style Ambidextrous
View More Specs

Products 1
Catalog Number Mfr. No. Length (Metric) Length (English) Size Price Quantity  
Catalog Number Mfr. No. Length (Metric) Length (English) Size Price Quantity  
19-010-266
View Documents
Ansell™
11251160W
40.6 cm 16 in. Wide
Case of 50 Each for $484.89
Only null left
 
Description

Description

  • Seamless design
  • Red and black over edge is to indicate wide (black) and narrow (red) for easy laundry sorting purpose
  • Special knitted bicep cuffs keeps sleeve in place
  • Available in different lengths and widths (narrow or wide) for optimal fit and comfort to fit every worker
  • Engineered to perform in combination with any HyFlex cut resistant glove
  • Useful in automotive and transport, aerospace, machinery and equipment, metal fabrication, white goods industries
Applications:
  • Adjusting panels parts components
  • Adjusting systems and screwing
  • Body Surface checking, marking and touching up
  • Glass repair and replacement
  • Handling metal sheets and panels
  • Handling parts with sharp, rough edges
  • Handling structural and body components
  • Inspection, selecting, checking parts
  • Press shop operations
  • Press shop operations
  • Seizing structural and body parts
Specifications

Specifications

Cut-resistant Sleeve
Dark Gray
Ambidextrous
Elastic Upper with Knit Cuff
Yes
Ansell INTERCEPT™ Technology
Non-sterile
Reusable
Seamless
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'