Learn More
Ansell™ HyFlex™ 11543
Gloves designed with INTERCEPT™ Cut Resistance Technology for improved workplace safety.
$2454.29 - $2454.29
Specifications
Construction Knit
Coating Material Nitrile
Color Black
Cut Resistance Rating ANSI A7
Hand Specific
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-010-489
View Documents
Ansell™
11543060
6
Case of 144 Pair for $2,454.29
Only null left
17-010-480
View Documents
Ansell™
11543070
7
Case of 144 Pair for $2,454.29
Only null left
17-010-487
View Documents
Ansell™
11543080
8
Case of 144 Pair for $2,454.29
Only null left
17-010-490
View Documents
Ansell™
11543090
9
Case of 144 Pair for $2,454.29
Only null left
17-010-481
View Documents
Ansell™
11543100
10
Case of 144 Pair for $2,454.29
Only null left
17-010-483
View Documents
Ansell™
11543110
11
Case of 144 Pair for $2,454.29
Only null left
Description
Description
Enhanced protection: Designed with INTERCEPT™ Cut Resistance Technology, these nitrile-coated gloves meet EN ISO F/ANSI A7 cut protection standards, for improved workplace safety.
  • Assured heat resistance: HyFlex™ 11-543 industrial safety gloves satisfy EN 407 contact heat level 1 requirements, enabling them to withstand heat up to 100°C or 212°F for at least 15 seconds
  • Improved durability: Featuring FORTIX™ Abrasion Resistance Technology, these HyFlex™ gloves offer excellent abrasion resistance than the previous generation, with a dirt-masking design further extending glove life
  • Reduced surface defect risks: HyFlex™ 11-543 industrial work gloves are also silicone-free, preventing contaminant transfer to metal before painting
Specifications
Specifications
Knit
Black
Specific
Cut-resistant Gloves
Knit
Straight
Nitrile
ANSI A7
Yes
Unisex
Lined
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'