missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell HyFlex™ 11-751 Medium Duty Abrasion-Resistant Gloves, INTERCEPT™ Technology
Level C cut protection gloves offer enhanced abrasion resistance for extended wear.
$155.62 - $1,867.48
Specifications
| ANSI Abrasion Rating | ANSI 6 |
|---|---|
| Certifications/Compliance (Europe) | CE, EN 420:2003 + A1:2009; Category II |
| Construction | Knit |
| Coating Material | Polyurethane |
| Color | Black |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19-050-082C
|
Ansell
11751080 |
8 |
N/A
|
|
|||||
|
19-050-082D
|
Ansell
11751090 |
9 |
N/A
|
|
|||||
|
19-050-082E
|
Ansell
11751100 |
10 |
N/A
|
|
|||||
|
19-050-082F
|
Ansell
11751110 |
11 |
N/A
|
|
|||||
Description
- INTERCEPT Technology™ for high level of cut protection
- High durability PU coating, increased levels of abrasion resistance
- Dirt masking design allows for extended product use
- In compliance with the standards EN420 (General requirements) and EN388 (Mechanical hazards)
- Also available in vendor pack
Specifications
| ANSI 6 | |
| Knit | |
| Black | |
| Specific | |
| HPPE, Fiberglass, Nylon, Spandex, Polyurethane | |
| EN 388:4X43C | |
| Knit | |
| 12 Pairs/Bag, 12 Bags/Cs. | |
| 8.8 in. |
| CE, EN 420:2003 + A1:2009; Category II | |
| Polyurethane | |
| ANSI A4 | |
| Yes | |
| Cut-resistant Gloves | |
| General maintenance, hardware assembly, machining, stamping, cutting, raw material handling | |
| HPPE, Fiberglass, Nylon, Spandex | |
| HyFlex™ 11-751 | |
| 22.4 cm |