Learn More
Ansell HyFlex™ 11-751 Medium Duty Abrasion-Resistant Gloves, INTERCEPT™ Technology
Level C cut protection gloves offer enhanced abrasion resistance for extended wear.
Supplier:  Ansell 11751100
 View more versions of this product
Catalog No. 19-050-082E
Only null left
Explore available promotions
Description
Description
  • INTERCEPT Technology™ for high level of cut protection
  • High durability PU coating, increased levels of abrasion resistance
  • Dirt masking design allows for extended product use
  • In compliance with the standards EN420 (General requirements) and EN388 (Mechanical hazards)
  • Also available in vendor pack
Specifications
Specifications
ANSI 6
Knit
Black
Specific
HPPE, Fiberglass, Nylon, Spandex, Polyurethane
10
General maintenance, hardware assembly, machining, stamping, cutting, raw material handling
HPPE, Fiberglass, Nylon, Spandex
12 Pairs/Bag, 12 Bags/Cs.
8.8 in.
CE, EN 420:2003 + A1:2009; Category II
Polyurethane
ANSI A4
Yes
Cut-resistant Gloves
EN 388:4X43C
Knit
11751100
HyFlex™ 11-751
22.4 cm
Product Suggestions
Product Suggestions