missing translation for 'onlineSavingsMsg'
Learn More

Medisca Hydroxyurea, USP

Catalog No. 14186239
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186239 25 g
14186162 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14186239 Supplier Medisca Supplier No. 135404*

Hydroxyurea USP is a white to off-white powder. It is somewhat hygroscopic, decomposing in the presence of moisture. Alternative name(s): hydroxycarbamide

Hydroxyurea USP is a white to off-white powder. It is somewhat hygroscopic, decomposing in the presence of moisture.

Alternative name(s): hydroxycarbamide

Specifications

Chemical Name or Material Hydroxyurea, USP
Packaging Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 127-07-1
Type Drug Standard
Molecular Weight (g/mol) 76.05
Quantity 25 g
Percent Purity 97 to 103 %
Grade USP