missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxypropyl Cellulose, 4,000-6,500 cP, NF, 53.4-850%, Spectrum™ Chemical
HY121, 9004-64-2
Supplier: Spectrum Chemical Mfg Cor HY12125KG
Description
Spectrum™ Chemical Hydroxypropyl Cellulose, 4,000-6,500 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 9004-64-2 | |
| 0.8% | |
| Poly Pail | |
| 2.5 kg | |
| 53.4 to 80.5% |
| 100% | |
| 5% | |
| (C24H44O16)n | |
| CC(O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)O)OC(-*)[C@H](OC(C)O)[C@H]2OC(C)O)[C@H](OC(C)O)[C@@H](OC(C)O)[C@@H]1O-* | |
| NF |
Chemical Identifiers
| 9004-64-2 | |
| (C24H44O16)n | |
| CC(O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)O)OC(-*)[C@H](OC(C)O)[C@H]2OC(C)O)[C@H](OC(C)O)[C@@H](OC(C)O)[C@@H]1O-* |