missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Cellulose, 4,000-6,500 cP, NF, 53.4-850%, Spectrum™ Chemical

Catalog No. p-7157598
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18603336 125 g Poly Bottle
18603337 500 g Poly Pail
18603338 2.5 kg Poly Pail
18603339 12 kg Fiber Drum
4 options

Catalog No. 18603336

Supplier: Spectrum Chemical Mfg Cor HY121125GM

Only null left

HY121, 9004-64-2

Spectrum™ Chemical Hydroxypropyl Cellulose, 4,000-6,500 cP, NF is a non-ionic, water soluble polymer used as a thickening agent and gelling agent for aqueous pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
SMILES CC(O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)O)OC(-*)[C@H](OC(C)O)[C@H]2OC(C)O)[C@H](OC(C)O)[C@@H](OC(C)O)[C@@H]1O-*

Specifications

CAS Max % 100%
Ignition Residue 0.8%
Loss on Drying 5%
Packaging Poly Bottle
Quantity 125 g
Percent Purity 53.4 to 80.5%
Grade NF