missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypivalic Acid 98.0+%, TCI America™

Catalog No. H066125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H066125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H066125G Supplier TCI America Supplier No. H066125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4835-90-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00059953
InChI Key RDFQSFOGKVZWKF-UHFFFAOYSA-N
Synonym hydroxypivalic acid, 3-hydroxypivalic acid, 2,2-dimethyl-3-hydroxypropionic acid, propanoic acid, 3-hydroxy-2,2-dimethyl, 3-hydroxy-2,2-dimethylpropionic acid, unii-bjp2cxk56p, bjp2cxk56p, acmc-1arbc, beta-hydroxy pivalic acid
PubChem CID 78548
IUPAC Name 3-hydroxy-2,2-dimethylpropanoic acid
SMILES CC(C)(CO)C(=O)O

Specifications

Melting Point 126°C
Color White
Quantity 25 g
Formula Weight 118.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hydroxypivalic Acid
EINECSNumber (2)-1351&(2)-1429
TSCA Yes