accessibility menu, dialog, popup

UserName

H3CCH3HOOHO

CAS RN 4835-90-9

IUPAC Name: 3-hydroxy-2,2-dimethylpropanoic acid
Synonyms: 3-hydroxy-2,2-dimethylpropanoic acid
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: RDFQSFOGKVZWKF-UHFFFAOYSA-N
SMILES: CC(C)(CO)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)