missing translation for 'onlineSavingsMsg'
Learn More

Hydroxynaphthol Blue, Dry in NaCl, Calcium and Hardness Indicator, ACS Grade, LabChem™

Supplier:  LabChem LC155707

 View more versions of this product

Catalog No. LC155707


Only null left

Description

Description

Specifications

Specifications

Hydroxynaphthol Blue
7647-14-5 , 63451-35-4
ClNa
MFCD00003477
Passes Test
[Na+].[Cl-]
58.44
CHEBI:26710
ACS
Passes Test
Purple
Granules
Dry in NaCl, Calcium and Hardness Indicator
99.25,0.75
C20H12N2Na2O11S3
Soluble in water
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
598.5
Poly Bottle
Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds; Hydrogen chloride
100 g

Chemical Identifiers

7647-14-5
58.44
FAPWRFPIFSIZLT-UHFFFAOYSA-M
CHEBI:26710
[Na+].[Cl-]
ClNa
MFCD00003477
5234
sodium chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions