missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine-O-sulfonic Acid 90.0+%, TCI America™

Catalog No. H053025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H053025G 25 g
H0530100G 100 g
H0530500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H053025G Supplier TCI America Supplier No. H053025G

Chemical Identifiers

CAS 2950-43-8
Molecular Formula H3NO4S
Molecular Weight (g/mol) 113.09
MDL Number MFCD00011604
InChI Key DQPBABKTKYNPMH-UHFFFAOYSA-N
Synonym hydroxylamine-o-sulfonic acid, aminooxy sulfonic acid, sulfoperamidic acid, hydroxylamine-o-sulphonic acid, permonosulfamic acid, amidoperoxymonosulfuric acid, aminooxysulfonic acid, amidosulfonic peracid, haos, sulfamic acid n-oxide
PubChem CID 76284
IUPAC Name (aminooxy)sulfonic acid
SMILES NOS(O)(=O)=O

Specifications

Color White to Yellow
Quantity 25 g
UN Number 3261
Formula Weight 113.09
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hydroxylamine-O-sulfonic Acid
EINECSNumber (2)-4012
RTECSNumber NC5427000
TSCA Yes