accessibility menu, dialog, popup

UserName

H2NOSOHOO

CAS RN 2950-43-8

IUPAC Name: (aminooxy)sulfonic acid
Synonyms: aminooxysulfonic acid
Molecular Weight (g/mol): 113.09
Molecular Formula: H3NO4S
InChi Key: DQPBABKTKYNPMH-UHFFFAOYSA-N
SMILES: NOS(O)(=O)=O
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (5)
  • (1)
  • (3)
  • (3)
  • (1)
  • (6)