missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine-O-sulfonic acid, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 210.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 0.97 |
| Packaging | Glass bottle |
| Linear Formula | H2NOSO3H |
| Quantity | 25 g |
| Fieser | 01,481; 02,217; 03,156; 04,256; 05,343; 06,290; 08,250; 09,245; 10,207; 15,17 |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 113.09 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Suspected of causing cancer. Causes serious eye damage. May cause an allergic skin reaction. Causes severe skin burns and eye damage. Harmful if swallowed. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear eye protection/face protection. Use personal protective equipment as required. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 220-971-6 |
| RTECSNumber | NC5427000 |
| TSCA | TSCA |
RUO – Research Use Only