Learn More
Hydroxylamine hydrochloride, 99%
CAS: 5470-11-1 | ClH4NO | 69.49 g/mol
Supplier: Thermo Scientific Chemicals A1539830
Description
Hydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.
Solubility
Soluble in water, alcohol, methanol and glycerol.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, phosphorous pentachloride, calcium and anhydrous copper(II) sulfate.
Specifications
| Hydroxylamine hydrochloride | |
| 5470-11-1 | |
| 250 g | |
| NH2OH·HCl | |
| UN2923 | |
| 14,4828 | |
| Soluble in water,alcohol,methanol and glycerol. | |
| Cl.NO | |
| 69.49 | |
| CHEBI:5807 | |
| 99% | |
| Hygroscopic |
| 152°C (dec.) | |
| 3.6 | |
| ClH4NO | |
| MFCD00051089 | |
| 3539763 | |
| hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
| WTDHULULXKLSOZ-UHFFFAOYSA-N | |
| hydroxylamine;hydrochloride | |
| 443297 | |
| 69.49 | |
| Acidic | |
| 1.67 |
Chemical Identifiers
| 5470-11-1 | |
| 69.49 | |
| WTDHULULXKLSOZ-UHFFFAOYSA-N | |
| 443297 | |
| hydroxylamine;hydrochloride |
| ClH4NO | |
| MFCD00051089 | |
| hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
| CHEBI:5807 | |
| Cl.NO |
Safety and Handling
GHS H Statement
H351-H373-H290-H302-H312-H317-H315-H319
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
May be corrosive to metals.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P234-P260-P264b-P270-P272-P280-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P390-P501c
H290-H301-H312-H315-H317-H319-H351-H373-H500
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber : 226-798-2
RTECSNumber : NC3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only