missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine hydrochloride, 99%

Catalog No. p-7022562
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
5000 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1539822 100 g
AAA1539830 250 g
AAA1539836 500 g
AAA153980B 1000 g
AAA153980I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AAA1539822 Supplier Thermo Scientific Chemicals Supplier No. A1539822
Only null left

CAS: 5470-11-1 | ClH4NO | 69.49 g/mol

Hydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.

Solubility
Soluble in water, alcohol, methanol and glycerol.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, phosphorous pentachloride, calcium and anhydrous copper(II) sulfate.

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine;hydrochloride
SMILES Cl.NO

Specifications

Melting Point 152°C (dec.)
pH 3.6
Quantity 100 g
Linear Formula NH2OH·HCl
UN Number UN2923
Beilstein 3539763
Merck Index 14,4828
Solubility Information Soluble in water,alcohol,methanol and glycerol.
Formula Weight 69.49
Percent Purity 99%
Odor Acidic
Sensitivity Hygroscopic
Density 1.67
Chemical Name or Material Hydroxylamine hydrochloride
Show More Show Less
Hazard Category H290-H301-H312-H315-H317-H319-H351-H373-H500
Hazard Statement GHS H Statement
H351-H373-H290-H302-H312-H317-H315-H319
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
May be corrosive to metals.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P234-P260-P264b-P270-P272-P280-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 226-798-2
RTECSNumber NC3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only