missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 10% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company R39010001C

Quantity 1 L
Packaging Amber Glass Bottle

Catalog No. R39010001C



Description

Description

Specifications

Chemical Identifiers

5470-11-1 , 7732-18-5
90.48, 9.52
3.2 (1.4% solution)
90.48, 9.52
1.05
Hydroxylamine Hydrochloride, 10% (w/v)

Specifications

Hydroxylamine Hydrochloride, 10% (w/v)
90.48, 9.52
ClH4NO
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
WTDHULULXKLSOZ-UHFFFAOYSA-N
hydroxylamine;hydrochloride
443297
Amber Glass Bottle
1 L
5470-11-1 , 7732-18-5
90.48, 9.52
1.05
Miscible in water
NO.Cl
69.49
CHEBI:5807
3.2 (1.4% solution)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Recommended Storage : Ambient Corrosive

SDS
Documents

Documents

Promotions

Promotions