missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyacetone, Technical

Catalog No. AC160341000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160341000 100 g Glass bottle
AC160345000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160341000 Supplier Thermo Scientific Chemicals Supplier No. 160341000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 116-09-6 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 116-09-6
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00004669
InChI Key XLSMFKSTNGKWQX-UHFFFAOYSA-N
Synonym hydroxyacetone, acetol, acetone alcohol, 1-hydroxy-2-propanone, 2-propanone, 1-hydroxy, 1-hydroxyacetone, methanol, acetyl, acetylmethanol, acetylcarbinol, 2-oxopropanol
PubChem CID 8299
ChEBI CHEBI:27957
IUPAC Name 1-hydroxypropan-2-one
SMILES CC(=O)CO

Specifications

Melting Point -17.0°C
Density 1.0800g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 56°C
Packaging Glass bottle
Refractive Index 1.4315
Linear Formula CH3COCH2OH
Quantity 100 g
UN Number 1224
Beilstein 01, 821
Specific Gravity 1.08
Merck Index 15, 64
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Viscosity 1.593 mPa.s (48°C)
Formula Weight 74.08
Grade Technical
Chemical Name or Material Hydroxyacetone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 204-124-8

RUO – Research Use Only