accessibility menu, dialog, popup

UserName

CH3OHO

CAS RN 116-09-6

IUPAC Name: 1-hydroxypropan-2-one
Synonyms: acetone alcohol pyruvic alcohol pyruvinalcohol methylketol
Molecular Weight (g/mol): 74.08
Molecular Formula: C3H6O2
InChi Key: XLSMFKSTNGKWQX-UHFFFAOYSA-N
SMILES: CC(=O)CO
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)