missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyacetone 80.0+%, TCI America™
$74.56 - $480.92
Chemical Identifiers
| CAS | 116-09-6 |
|---|---|
| Molecular Formula | C3H6O2 |
| Molecular Weight (g/mol) | 74.079 |
| MDL Number | MFCD00004669 |
| InChI Key | XLSMFKSTNGKWQX-UHFFFAOYSA-N |
| Synonym | hydroxyacetone, acetol, acetone alcohol, 1-hydroxy-2-propanone, 2-propanone, 1-hydroxy, 1-hydroxyacetone, methanol, acetyl, acetylmethanol, acetylcarbinol, 2-oxopropanol |
| PubChem CID | 8299 |
| ChEBI | CHEBI:27957 |
| IUPAC Name | 1-hydroxypropan-2-one |
| SMILES | CC(=O)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H038825ML
|
TCI America
H038825ML |
25 mL |
Each for $74.56
|
|
|||||
|
H0388100ML
|
TCI America
H0388100ML |
100 mL |
Each for $126.76
|
|
|||||
|
H0388500ML
|
TCI America
H0388500ML |
500 mL |
Each for $480.92
|
|
|||||
Chemical Identifiers
| 116-09-6 | |
| 74.079 | |
| XLSMFKSTNGKWQX-UHFFFAOYSA-N | |
| 8299 | |
| 1-hydroxypropan-2-one |
| C3H6O2 | |
| MFCD00004669 | |
| hydroxyacetone, acetol, acetone alcohol, 1-hydroxy-2-propanone, 2-propanone, 1-hydroxy, 1-hydroxyacetone, methanol, acetyl, acetylmethanol, acetylcarbinol, 2-oxopropanol | |
| CHEBI:27957 | |
| CC(=O)CO |
Specifications
| 116-09-6 | |
| C3H6O2 | |
| 25 mL | |
| hydroxyacetone, acetol, acetone alcohol, 1-hydroxy-2-propanone, 2-propanone, 1-hydroxy, 1-hydroxyacetone, methanol, acetyl, acetylmethanol, acetylcarbinol, 2-oxopropanol | |
| CC(=O)CO | |
| 74.079 | |
| CHEBI:27957 | |
| ≥80.0% (GC) | |
| Hydroxyacetone |
| Yellow | |
| MFCD00004669 | |
| 1993 | |
| XLSMFKSTNGKWQX-UHFFFAOYSA-N | |
| 1-hydroxypropan-2-one | |
| 8299 | |
| 74.08 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3205
RTECSNumber : UC2800000
TSCA : Yes
Recommended Storage : Refrigerator